Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALG10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560328
Description
Immunogen sequence: SRALREPYMD EIFHLPQAQR YCEGHFSLSQ WDPMITTLP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
ALG10 encodes a membrane-associated protein that adds the third glucose residue to the lipid-linked oligosaccharide precursor for N-linked glycosylation. That is, it transfers the terminal glucose from dolichyl phosphate glucose (Dol-P-Glc) onto the lipid-linked oligosaccharide Glc2Man9GlcNAc(2)-PP-Dol. The rat protein homolog was shown to specifically modulate the gating function of the rat neuronal ether-a-go-go (EAG) potassium ion channel.
Specifications
ALG10 | |
Polyclonal | |
Unconjugated | |
ALG10 | |
ALG10; ALG10, alpha-1,2-glucosyltransferase; ALG10A; Alpha-1,2-glucosyltransferase ALG10-A; alpha2-glucosyltransferase; alpha-2-glucosyltransferase ALG10-A; asparagine-linked glycosylation 10 homolog (yeast, alpha-1,2-glucosyltransferase); asparagine-linked glycosylation 10, alpha-1,2-glucosyltransferase homolog; asparagine-linked glycosylation protein 10 homolog A; derepression of ITR1 expression 2 homolog; DIE2; dolichyl-P-Glc:Glc(2)Man(9)GlcNAc(2)-PP-dolichol alpha-1,2- glucosyltransferase; Dol-P-Glc:Glc(2)Man(9)GlcNAc(2)-PP-Dol alpha-1,2-glucosyltransferase; KCR1; potassium channel regulator 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5BKT4 | |
ALG10 | |
Recombinant protein corresponding to Human ALG10. Recombinant protein control fragment (Product #RP-99517). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction