Learn More
Invitrogen™ ALG10B Polyclonal Antibody

Description
ALG10B Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | ALG10B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ALG10B |
| Gene Accession No. | Q5I7T1 |
| Gene Alias | ALG10; ALG10B; ALG10B, alpha-1,2-glucosyltransferase; alpha-1,2-glucosyltransferase ALG10-A; Alpha-2-glucosyltransferase ALG10-B; asparagine-linked glycosylation 10 homolog B (yeast, alpha-1,2-glucosyltransferase); asparagine-linked glycosylation 10, alpha-1,2-glucosyltransferase homolog B; asparagine-linked glycosylation protein 10 homolog B; dolichyl-P-Glc:Glc(2)Man(9)GlcNAc(2)-PP-dolichol alpha-1,2- glucosyltransferase; KCR1; modifier of the HERG potassium channel; potas; potassium channel regulator 1; putative alpha-1,2-glucosyltransferase ALG10-B; putative Dol-P-Glc:Glc(2)Man(9)GlcNAc(2)-PP-Dol alpha-1,2-glucosyltransferase |
| Gene Symbols | ALG10B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.