Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542578
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542578 Supplier Invitrogen™ Supplier No. PA542578
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LHTMWNEHFG IGVVECMAAG TIILAHNSGG PKLDIVIPHE GDITGFLAES Sequence homology: Cow: 92%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 93%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Yeast: 92%; Zebrafish: 91%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG11
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ALG11
Gene Accession No. Q2TAA5
Gene Alias AI849156; Alg11; ALG11 alpha-1,2-mannosyltransferase; ALG11, alpha-1,2-mannosyltransferase; asparagine-linked glycosylation 11 (alpha-1,2-mannosyltransferase); asparagine-linked glycosylation 11 homolog (S. cerevisiae, alpha-1,2-mannosyltransferase); asparagine-linked glycosylation 11 homolog (yeast, alpha-1,2-mannosyltransferase); asparagine-linked glycosylation 11, alpha-1,2-mannosyltransferase homolog; asparagine-linked glycosylation protein 11 homolog; AW492253; B230397C21; CDG1P; defective mannosyltransferase IV(V); fj04e04; GDP-Man:Man(3)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase; GDP-Man:Man(3)GlcNAc(2)-PP-dolichol alpha-1,2-mannosyltransferase; glycolipid 2-alpha-mannosyltransferase; GT8; LOC407614 protein; mannosyltransferase IV(V); Mmat-4(5); RGD1564725; si:dkey-1h24.5; wu:fj04e04
Gene Symbols ALG11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ALG11.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440138
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.