Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG11 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988159
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8159 Supplier Bioss Supplier No. BS7957R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function remains unknown.

Specifications

Antigen ALG11
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ALG11
Gene Accession No. Q2TAA5
Gene Alias AI849156; ALG11; ALG11 alpha-1,2-mannosyltransferase; ALG11, alpha-1,2-mannosyltransferase; Alpha-1,2-mannosyltransferase ALG11; asparagine-linked glycosylation 11 (alpha-1,2-mannosyltransferase); asparagine-linked glycosylation 11 homolog (S. cerevisiae, alpha-1,2-mannosyltransferase); asparagine-linked glycosylation 11 homolog (yeast, alpha-1,2-mannosyltransferase); asparagine-linked glycosylation 11, alpha-1,2-mannosyltransferase homolog; asparagine-linked glycosylation 11, alpha-1,2-mannosyltransferase homolog (yeast); Asparagine-linked glycosylation protein 11; Asparagine-linked glycosylation protein 11 homolog; AW492253; B230397C21; BOS_12437; CDG1P; defective mannosyltransferase IV(V); fj04e04; GDP-Man:Man(3)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase; GDP-Man:Man(3)GlcNAc(2)-PP-dolichol alpha-1,2-mannosyltransferase; Glycolipid 2-alpha-mannosyltransferase; GT8; LOC407614 protein; mannosyltransferase IV(V); Mmat-4(5); N2510; RGD1564725; si:dkey-1h24.5; wu:fj04e04; YNL048W; YNL2510W
Gene Symbols ALG11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ALG11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440138
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.