Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALG12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ALG12 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ALG12 Polyclonal specifically detects ALG12 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ALG12 | |
Polyclonal | |
Rabbit | |
Human | |
79087 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KVEESFNLQATHDLLYHWQDLEQYDHLEF | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
alpha-1,6-mannosyltransferase), asparagine-linked glycosylation 12 homolog (S. cerevisiae, asparagine-linked glycosylation 12 homolog (yeast, asparagine-linked glycosylation 12, alpha-1,6-mannosyltransferase homolog (S.cerevisiae), Asparagine-linked glycosylation protein 12 homolog, CDG1G, dolichyl-P-Man:Man(7)GlcNAc(2)-PP-dolichyl-alpha-1,6-mannosyltransferase, dolichyl-P-mannose:Man-7-GlcNAc-2-PP-dolichyl-alpha-6-mannosyltransferase, EC 2.4.1.-, ECM39, hALG12, Mannosyltransferase ALG12 homolog, Membrane protein SB87, MGC111358, MGC3136, PP14673 | |
ALG12 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title