Learn More
Invitrogen™ ALG12 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ALG12 protein.
Specifications
Specifications
| Antigen | ALG12 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ALG12 |
| Gene Accession No. | Q9BV10 |
| Gene Alias | ALG12; ALG12, alpha-1,6-mannosyltransferase; asparagine-linked glycosylation 12 homolog (S. cerevisiae, alpha-1,6-mannosyltransferase); asparagine-linked glycosylation 12 homolog (yeast, alpha-1,6-mannosyltransferase); asparagine-linked glycosylation 12, alpha-1,6-mannosyltransferase homolog; asparagine-linked glycosylation protein 12 homolog; CDG1G; dolichyl-P-Man:Man(7)GlcNAc(2)-PP-dolichol alpha-1,6-mannosyltransferase; dolichyl-P-Man:Man(7)GlcNAc(2)-PP-dolichyl-alpha-1,6-mannosyltransferase; dolichyl-P-mannose:Man-7-GlcNAc-2-PP-dolichyl-alpha-6-mannosyltransferase; dol-P-Man dependent alpha-1,6-mannosyltransferase; dol-P-Man:Man(7)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; ECM39; hALG12; Mannosyltransferase ALG12 homolog; membrane protein SB87; PP14673 |
| Gene Symbols | ALG12 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.