Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562452
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562452 Supplier Invitrogen™ Supplier No. PA562452
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGGVAMQRLH QLVPPQTDVL LHIDVAAAQT GVSRFLQVNS AWRYDKREDV QPGTGMLAYT HILMEAAPGL LALYRDTHRV LASVVGTTGV SLNLTQLPPF NVHLQTKLVL LERL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.

This gene encodes a member of the glycosyltransferase 22 family. The encoded protein catalyzes the addition of the eighth mannose residue in an alpha-1,6 linkage onto the dolichol-PP-oligosaccharide precursor (dolichol-PP-Man(7)GlcNAc(2)) required for protein glycosylation. Mutations in this gene have been associated with congenital disorder of glycosylation type Ig (CDG-Ig)characterized by abnormal N-glycosylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALG12
Gene Accession No. Q9BV10
Gene Alias ALG12; ALG12, alpha-1,6-mannosyltransferase; asparagine-linked glycosylation 12 homolog (S. cerevisiae, alpha-1,6-mannosyltransferase); asparagine-linked glycosylation 12 homolog (yeast, alpha-1,6-mannosyltransferase); asparagine-linked glycosylation 12, alpha-1,6-mannosyltransferase homolog; asparagine-linked glycosylation protein 12 homolog; CDG1G; dolichyl-P-Man:Man(7)GlcNAc(2)-PP-dolichol alpha-1,6-mannosyltransferase; dolichyl-P-Man:Man(7)GlcNAc(2)-PP-dolichyl-alpha-1,6-mannosyltransferase; dolichyl-P-mannose:Man-7-GlcNAc-2-PP-dolichyl-alpha-6-mannosyltransferase; dol-P-Man dependent alpha-1,6-mannosyltransferase; dol-P-Man:Man(7)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; ECM39; hALG12; Mannosyltransferase ALG12 homolog; membrane protein SB87; PP14673
Gene Symbols ALG12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALG12. Recombinant protein control fragment (Product #RP-91538).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79087
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.