Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALG13 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP320219AF532
Description
ALG13 Polyclonal antibody specifically detects ALG13 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ALG13 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Asparagine-linked glycosylation 13 homolog, asparagine-linked glycosylation 13 homolog (S. cerevisiae), chromosome X open reading frame 45, CXorf45, EC 2.4.1.141, FLJ23018, FLJ31785, GLT28D1, glycosyltransferase 28 domain containing 1, Glycosyltransferase 28 domain-containing protein 1, hematopoietic stem/progenitor cells protein MDS031, MDS031, MGC12423, UDP-N-acetylglucosamine transferase subunit ALG13 homolog, YGL047W | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ALG13 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
79868 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction