Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG13 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB274795 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274795 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274795 Supplier Novus Biologicals Supplier No. NBP320219F
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALG13 Polyclonal antibody specifically detects ALG13 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ALG13
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Asparagine-linked glycosylation 13 homolog, asparagine-linked glycosylation 13 homolog (S. cerevisiae), chromosome X open reading frame 45, CXorf45, EC 2.4.1.141, FLJ23018, FLJ31785, GLT28D1, glycosyltransferase 28 domain containing 1, Glycosyltransferase 28 domain-containing protein 1, hematopoietic stem/progenitor cells protein MDS031, MDS031, MGC12423, UDP-N-acetylglucosamine transferase subunit ALG13 homolog, YGL047W
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ALG13
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79868
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.