Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551754 Supplier Invitrogen™ Supplier No. PA551754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGTTSFDDLI ACVSAPDSLQ KIESLGYNRL ILQIGRGTVV PEPFSTESFT LDVYRYKDSL KEDIQKADLV ISHAGAGSCL ETLEKGKPLV VVINEKLMNN HQLELAKQLH KEGHLFYCTC STLPGLLQSM DLSTLKCYPP G Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

Isoform 1: Possible multifunctional enzyme with both glycosyltransferase and deubiquitinase activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALG13
Gene Accession No. Q9NP73
Gene Alias 2810046O15Rik; 4833435D08Rik; Alg13; ALG13, UDP-N-acetylglucosaminyltransferase subunit; asparagine-linked glycosylation 13; asparagine-linked glycosylation 13 homolog; CDG1S; CXorf45; EIEE36; Glt28d1; glycosyltransferase 28 domain containing 1; glycosyltransferase 28 domain-containing protein 1; hematopoietic stem/progenitor cells protein MDS031; MDS031; N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase; OTTMUSG00000018917; Putative bifunctional UDP-N-acetylglucosamine transferase and deubiquitinase ALG13; TDRD13; tudor domain containing 13; UDP-N-acetylglucosamine transferase subunit ALG13 homolog; YGL047W
Gene Symbols ALG13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALG13. Recombinant protein control fragment (Product #RP-92169).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79868
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.