Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116801 Supplier Invitrogen™ Supplier No. PA5116801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Raji, K-562, Jurkat, HepG2, Mouse liver, Mouse kidney.

Isoform 1: Possible multifunctional enzyme with both glycosyltransferase and deubiquitinase activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG13
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ALG13
Gene Accession No. Q9D8C3, Q9NP73
Gene Alias 2810046O15Rik; 4833435D08Rik; Alg13; ALG13, UDP-N-acetylglucosaminyltransferase subunit; asparagine-linked glycosylation 13; asparagine-linked glycosylation 13 homolog; CDG1S; CXorf45; EIEE36; Glt28d1; glycosyltransferase 28 domain containing 1; glycosyltransferase 28 domain-containing protein 1; hematopoietic stem/progenitor cells protein MDS031; MDS031; N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase; OTTMUSG00000018917; Putative bifunctional UDP-N-acetylglucosamine transferase and deubiquitinase ALG13; TDRD13; tudor domain containing 13; UDP-N-acetylglucosamine transferase subunit ALG13 homolog; YGL047W
Gene Symbols ALG13
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67574, 79868
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.