Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG2 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
SDP

Catalog No. 501674944
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4944 100 μL
50-254-0447 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-4944 Supplier OriGene Supplier No. TA504215
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALG2 is a member of the glycosyltransferase 1 family. It acts as an alpha 1, 3 mannosyltransferase, mannosylating Man(2)GlcNAc(2)-dolichol diphosphate and Man(1)GlcNAc(2)-dolichol diphosphate to form Man(3)GlcNAc(2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii).This gene encodes a member of the glycosyltransferase 1 family. The encoded protein acts as an alpha 1, 3 mannosyltransferase, mannosylating Man(2)GlcNAc(2)-dolichol diphosphate and Man(1)GlcNAc(2)-dolichol diphosphate to form Man(3)GlcNAc(2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii).

Specifications

Antigen ALG2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ALG2
Gene Accession No. Q9H553
Gene Alias CDG1I, CDGIi, CMS14, CMSTA3, NET38, hALPG2
Gene Symbols ALG2
Host Species Mouse
Immunogen Full length human recombit protein of human ALG2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85365
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.