Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG2 Monoclonal Antibody (OTI1E7), TrueMAB™, OriGene
SDP

Catalog No. 502540445
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0445 30 μL
50-167-4946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0445 Supplier OriGene Supplier No. TA504218S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALG2 Monoclonal antibody specifically detects ALG2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ALG2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E7
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ALG2
Gene Accession No. Q9H553
Gene Alias 1110018A23Rik; 1300013N08Rik; Alg2; ALG2 alpha-1,3/1,6-mannosyltransferase; ALG2, alpha-1,3/1,6-mannosyltransferase; ALPG2; alpha-1,3/1,6-mannosyltransferase ALG2; alpha-1,3-mannosyltransferase ALG2; asparagine-linked glycosylation 2 (alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (S. cerevisiae, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (yeast, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog; asparagine-linked glycosylation protein 2 homolog; CDGIi; CMS14; CMSTA3; FLJ14511; FLJ14511 protein; GDP-Man:Man(1)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase; GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase; GDP-Man:Man(2)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; hALPG2; homolog of yeast ALG2; im:7145131; LOW QUALITY PROTEIN: alpha-1,3/1,6-mannosyltransferase ALG2; MNCb-5081; NET38; UNQ666/PRO1298
Host Species Mouse
Immunogen Full length human recombinant protein of human ALG2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85365
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.