Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALG2 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504217S


Description
ALG2 Monoclonal antibody specifically detects ALG2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
ALG2 | |
Monoclonal | |
0.83 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H553 | |
Mouse | |
Affinity chromatography | |
RUO | |
85365 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2A3 | |
Unconjugated | |
ALG2 | |
1110018A23Rik; 1300013N08Rik; Alg2; ALG2 alpha-1,3/1,6-mannosyltransferase; ALG2, alpha-1,3/1,6-mannosyltransferase; ALPG2; alpha-1,3/1,6-mannosyltransferase ALG2; alpha-1,3-mannosyltransferase ALG2; asparagine-linked glycosylation 2 (alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (S. cerevisiae, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (yeast, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog; asparagine-linked glycosylation protein 2 homolog; CDGIi; CMS14; CMSTA3; FLJ14511; FLJ14511 protein; GDP-Man:Man(1)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase; GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase; GDP-Man:Man(2)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; hALPG2; homolog of yeast ALG2; im:7145131; LOW QUALITY PROTEIN: alpha-1,3/1,6-mannosyltransferase ALG2; MNCb-5081; NET38; UNQ666/PRO1298 | |
Full length human recombinant protein of human ALG2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction