Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559643 Supplier Invitrogen™ Supplier No. PA559643
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFLLLSINRY ERKKNLTLAL EALVQLRGRL TSQDWERVHL IVAGGYDERV LENVEHYQEL KKMVQQSDLG QYVTFLRSFS DKQKISLLH Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.

ALG2 is a member of the glycosyltransferase 1 family. It acts as an alpha 1, 3 mannosyltransferase, mannosylating Man(2)GlcNAc(2)-dolichol diphosphate and Man(1)GlcNAc(2)-dolichol diphosphate to form Man(3)GlcNAc(2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii).This gene encodes a member of the glycosyltransferase 1 family. The encoded protein acts as an alpha 1, 3 mannosyltransferase, mannosylating Man(2)GlcNAc(2)-dolichol diphosphate and Man(1)GlcNAc(2)-dolichol diphosphate to form Man(3)GlcNAc(2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALG2
Gene Accession No. Q9H553
Gene Alias 1110018A23Rik; 1300013N08Rik; Alg2; ALG2 alpha-1,3/1,6-mannosyltransferase; ALG2, alpha-1,3/1,6-mannosyltransferase; ALPG2; alpha-1,3/1,6-mannosyltransferase ALG2; alpha-1,3-mannosyltransferase ALG2; asparagine-linked glycosylation 2 (alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (S. cerevisiae, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (yeast, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog; asparagine-linked glycosylation protein 2 homolog; CDGIi; CMS14; CMSTA3; FLJ14511; FLJ14511 protein; GDP-Man:Man(1)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase; GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase; GDP-Man:Man(2)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; hALPG2; homolog of yeast ALG2; im:7145131; LOW QUALITY PROTEIN: alpha-1,3/1,6-mannosyltransferase ALG2; MNCb-5081; NET38; UNQ666/PRO1298
Gene Symbols ALG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALG2. Recombinant protein control fragment (Product #RP-98410).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85365
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.