Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543263 Supplier Invitrogen™ Supplier No. PA543263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QSDLGQYVTF LRSFSDKQKI SLLHSCTCVL YTPSNEHFGI VPLEAMYMQC Sequence homology: Cow: 100%; Dog: 93%; Goat: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 93%.

ALG2 is a member of the glycosyltransferase 1 family. It acts as an alpha 1, 3 mannosyltransferase, mannosylating Man(2)GlcNAc(2)-dolichol diphosphate and Man(1)GlcNAc(2)-dolichol diphosphate to form Man(3)GlcNAc(2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii).This gene encodes a member of the glycosyltransferase 1 family. The encoded protein acts as an alpha 1, 3 mannosyltransferase, mannosylating Man(2)GlcNAc(2)-dolichol diphosphate and Man(1)GlcNAc(2)-dolichol diphosphate to form Man(3)GlcNAc(2)-dolichol diphosphate. Defects in this gene have been associated with congenital disorder of glycosylation type Ih (CDG-Ii).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ALG2
Gene Accession No. Q9H553
Gene Alias 1110018A23Rik; 1300013N08Rik; Alg2; ALG2 alpha-1,3/1,6-mannosyltransferase; ALG2, alpha-1,3/1,6-mannosyltransferase; ALPG2; alpha-1,3/1,6-mannosyltransferase ALG2; alpha-1,3-mannosyltransferase ALG2; asparagine-linked glycosylation 2 (alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (S. cerevisiae, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (yeast, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog; asparagine-linked glycosylation protein 2 homolog; CDGIi; CMS14; CMSTA3; FLJ14511; FLJ14511 protein; GDP-Man:Man(1)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase; GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase; GDP-Man:Man(2)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; hALPG2; homolog of yeast ALG2; im:7145131; LOW QUALITY PROTEIN: alpha-1,3/1,6-mannosyltransferase ALG2; MNCb-5081; NET38; UNQ666/PRO1298
Gene Symbols ALG2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ALG2 (aa 290-339).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85365
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.