Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552496 Supplier Invitrogen™ Supplier No. PA552496
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RHEEEKFFLN AKGQKETLPS IWDSPTKQLS VVVPSYNEEK RLPVMMDEAL SYLEKRQKRD PAFTYEVIVV DDGSKDQTSK VAFKYCQKYG SDKVRVITLV KNRGKGGAIR MGIFSSRGEK ILMADADGAT KFPDVEKLEK GLNDL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Dolichyl-phosphate beta-glucosyltransferase (ALG5), belongs to the family of glycosyltransferases, specifically the hexosyltransferases. ALG5 is widely expressed in pancreas, placenta, liver, heart, brain, kidney, skeletal muscle, and lung. ALG5 can catalyze the chemical reaction between UDP-glucose and UDP with dolichyl phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Alg5
Gene Accession No. Q9DB25, Q9Y673
Gene Alias 1500026A19Rik; 2600005J22Rik; ALG5; Alg5 homolog; ALG5, dolichyl-phosphate beta-glucosyltransferase; Alg5, S. cerevisiae, homolog of; asparagine-linked glycosylation 5 (dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5 homolog (S. cerevisiae, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5 homolog (yeast, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5, dolichyl-phosphate beta-glucosyltransferase homolog; asparagine-linked glycosylation protein 5 homolog; AV009521; AV170001; AW545564; bA421P11.2; dolichyl phosphate glucosyltransferase; dolichyl-phosphate beta-glucosyltransferase; DolP-glucosyltransferase; HSPC149
Gene Symbols Alg5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALG5. Recombinant protein control fragment (Product #RP-90158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295051, 29880, 66248
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.