Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG6 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990124
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0124 25 μL
50-199-0123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0124 Supplier Rockland Immunochemicals Supplier No. 100401GJ0S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. ALG6 Antibody antiserum is directed against human ALG6. A 58 kDa band is detected when assayed by immunoblot. Cross reactivity is expected against human ALG6 protein based on a 100% homology of the amino acid sequence of the immunogen between rat, mouse, drosophila, and pongo abelii.

ALG6 is a member of the ALG6/ALG8 glucosyltransferase family. It catalyzes the addition of the first glucose residue to the growing lipid-linked oligosaccharide precursor of N-linked glycosylation. Mutations in this gene encoding ALG6 are associated with congenital disorders of glycosylation type Ic.This gene encodes a member of the ALG6/ALG8 glucosyltransferase family. The encoded protein catalyzes the addition of the first glucose residue to the growing lipid-linked oligosaccharide precursor of N-linked glycosylation. Mutations in this gene are associated with congenital disorders of glycosylation type Ic.

Specifications

Antigen ALG6
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 80 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ALG6
Gene Accession No. Q9Y672
Gene Alias ALG6; ALG6 alpha-1,3-glucosyltransferase; ALG6, alpha-1,3-glucosyltransferase; asparagine-linked glycosylation 6 (alpha-1,3,-glucosyltransferase); asparagine-linked glycosylation 6 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 6 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase)-like; asparagine-linked glycosylation 6 homolog (yeast, alpha-1,3,-glucosyltransferase); asparagine-linked glycosylation 6 homolog (yeast, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 6, alpha-1,3-glucosyltransferase homolog; asparagine-linked glycosylation 6, alpha-1,3-glucosyltransferase homolog (S. cerevisiae); Asparagine-linked glycosylation protein 6; Asparagine-linked glycosylation protein 6 homolog; BOS_3497; CDG1C; Dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase; dolichyl-P-Glc:Man(9)GlcNAc(2)-PP-dolichol alpha- 1->3-glucosyltransferase; dolichyl-P-Glc:Man(9)GlcNAc(2)-PP-dolichol alpha-1,3-glucosyltransferase; Dolichyl-P-Glc:Man9GlcNAc2-PP-dolichyl glucosyltransferase; dolichyl-P-Glc:Man9GlcNAc2-PP-dolichylglucosyltransferase; Dol-P-Glc:Man(9)GlcNAc(2)-PP-Dol alpha-1,3-glucosyltransferase; E230028F23Rik; LOC100357591; Man(9)GlcNAc(2)-PP-Dol alpha-1,3-glucosyltransferase; My046; UNA544; YOR002W; zgc:100813
Gene Symbols ALG6
Host Species Rabbit
Immunogen This whole rabbit serum was prepared by repeated immunizations with a synthetic peptide, corresponding to a region near the N-terminus of human ALG6 protein, conjugated to KLH.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29929
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.