Learn More
Invitrogen™ ALG8 Monoclonal Antibody (2E10)

Description
Peptide Sequence: NQLPQVFSRL FPFKRGLCHA YWAPNFWALY NALDKVLSVI GLKLKFLDPN NIPKASMTSG LVQQFQHTVL PSVTP.
Specifications
Specifications
| Antigen | ALG8 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2E10 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ALG8 |
| Gene Accession No. | Q9BVK2 |
| Gene Alias | AI447372; alg8; ALG8 alpha-1,3-glucosyltransferase; ALG8, alpha-1,3-glucosyltransferase; asparagine-linked glycosylation 8 (alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8 alpha-13-glucosyltransferase-like protein; asparagine-linked glycosylation 8 homolog; asparagine-linked glycosylation 8 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8 homolog (yeast, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8, alpha-1,3-glucosyltransferase homolog; asparagine-linked glycosylation protein 8 homolog; CDG1H; dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase; dolichyl-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichol alpha- 1->3-glucosyltransferase; dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase; dolichyl-P-glucose:Glc1Man9GlcNAc2-PP-dolichyl-alpha-1,3-glucosyltransferase; Dol-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichyl alpha-1,3-glucosyltransferase; Gm1089; HUSSY-02; hypothetical protein LOC327601; probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase; putative dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase-like protein; si:rp71-1p14.2; Unknown (protein for MGC:142391); wu:fi19e09; wu:fi19f07; zgc:112507 |
| Gene Symbols | ALG8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.