Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550317 Supplier Invitrogen™ Supplier No. PA550317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ALG8 protein.

Adds the second glucose residue to the lipid-linked oligosaccharide precursor for N-linked glycosylation. Transfers glucose from dolichyl phosphate glucose (Dol-P-Glc) onto the lipid-linked oligosaccharide Glc(1)Man(9)GlcNAc(2)-PP-Dol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ALG8
Gene Accession No. Q9BVK2
Gene Alias AI447372; alg8; ALG8 alpha-1,3-glucosyltransferase; ALG8, alpha-1,3-glucosyltransferase; asparagine-linked glycosylation 8 (alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8 alpha-13-glucosyltransferase-like protein; asparagine-linked glycosylation 8 homolog; asparagine-linked glycosylation 8 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8 homolog (yeast, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8, alpha-1,3-glucosyltransferase homolog; asparagine-linked glycosylation protein 8 homolog; CDG1H; dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase; dolichyl-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichol alpha- 1->3-glucosyltransferase; dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase; dolichyl-P-glucose:Glc1Man9GlcNAc2-PP-dolichyl-alpha-1,3-glucosyltransferase; Dol-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichyl alpha-1,3-glucosyltransferase; Gm1089; HUSSY-02; hypothetical protein LOC327601; probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase; putative dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase-like protein; si:rp71-1p14.2; Unknown (protein for MGC:142391); wu:fi19e09; wu:fi19f07; zgc:112507
Gene Symbols ALG8
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human ALG8, alpha-1,3-glucosyltransferase.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79053
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.