Learn More
Invitrogen™ ALG9 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ALG9 protein.
Specifications
Specifications
| Antigen | ALG9 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ALG9 |
| Gene Accession No. | Q9H6U8 |
| Gene Alias | ALG9; ALG9, alpha-1,2-mannosyltransferase; alpha-1,2-mannosyltransferase ALG9; asparagine-linked glycosylation 9 alpha-12-mannosyltransferase-like protein; asparagine-linked glycosylation 9 homolog (S. cerevisiae, alpha- 1,2-mannosyltransferase); asparagine-linked glycosylation 9 homolog (yeast, alpha- 1,2-mannosyltransferase); asparagine-linked glycosylation 9, alpha-1,2-mannosyltransferase homolog; asparagine-linked glycosylation protein 9 homolog; CDG1L; DIBD1; disrupted in bipolar affective disorder 1; disrupted in bipolar disorder protein 1; dolichyl-P-Man:Man(6)GlcNAc(2)-PP-dolichol alpha-1,2-mannosyltransferase; dolichyl-P-Man:Man(8)GlcNAc(2)-PP-dolichol alpha-1,2-mannosyltransferase; dol-P-Man dependent alpha-1,2-mannosyltransferase; dol-P-Man:Man(6)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase; Dol-P-Man:Man(8)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase; GIKANIS; LOH11CR1J; loss of heterozygosity, 11, chromosomal region 1 gene J product |
| Gene Symbols | ALG9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.