Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558447
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558447 Supplier Invitrogen™ Supplier No. PA558447
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALFRGYHGPL DLYPEFYRIA TDPTIHTVPE GRPVNVCVGK EWYRFPSSFL LPDNWQLQFI PSEFRGQLPK PFAEGPLATR IVPTDMNDQN L Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 24%.

This gene encodes an alpha-1,2-mannosyltransferase enzyme that functions in lipid-linked oligosaccharide assembly. Mutations in this gene result in congenital disorder of glycosylation type Il. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALG9
Gene Accession No. Q9H6U8
Gene Alias ALG9; ALG9, alpha-1,2-mannosyltransferase; alpha-1,2-mannosyltransferase ALG9; asparagine-linked glycosylation 9 alpha-12-mannosyltransferase-like protein; asparagine-linked glycosylation 9 homolog (S. cerevisiae, alpha- 1,2-mannosyltransferase); asparagine-linked glycosylation 9 homolog (yeast, alpha- 1,2-mannosyltransferase); asparagine-linked glycosylation 9, alpha-1,2-mannosyltransferase homolog; asparagine-linked glycosylation protein 9 homolog; CDG1L; DIBD1; disrupted in bipolar affective disorder 1; disrupted in bipolar disorder protein 1; dolichyl-P-Man:Man(6)GlcNAc(2)-PP-dolichol alpha-1,2-mannosyltransferase; dolichyl-P-Man:Man(8)GlcNAc(2)-PP-dolichol alpha-1,2-mannosyltransferase; dol-P-Man dependent alpha-1,2-mannosyltransferase; dol-P-Man:Man(6)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase; Dol-P-Man:Man(8)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase; GIKANIS; LOH11CR1J; loss of heterozygosity, 11, chromosomal region 1 gene J product
Gene Symbols ALG9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALG9. Recombinant protein control fragment (Product #RP-97282).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79796
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.