Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alginate Antibody (3G4-1F5), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259349C
Description
Alginate Monoclonal specifically detects Alginate in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Alginate | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
Primary | |
Non-species specific | |
Purified |
Western Blot, Immunohistochemistry | |
3G4-1F5 | |
Western Blot, Immunohistochemistry | |
Gum levan, Kelacid, Levan gum, Sazio | |
Sodium Alginate conjugated to KLH | |
100 μL | |
Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction