Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alginate Antibody (3G4-1F5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259349F
Description
Alginate Monoclonal specifically detects Alginate in Non-species specific samples. It is validated for Immunohistochemistry.Specifications
| Alginate | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Non-species specific | |
| Purified |
| Immunohistochemistry | |
| 3G4-1F5 | |
| Immunohistochemistry | |
| Gum levan, Kelacid, Levan gum, Sazio | |
| Sodium Alginate conjugated to KLH | |
| 100 μL | |
| Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction