Learn More
Alginate Antibody (3G4-1F5), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259349H
Catalog No. NBP259349H
Only null left
Add to Cart
Description
Description
Alginate Monoclonal specifically detects Alginate in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
Alginate
Monoclonal
HRP
PBS with No Preservative
Mouse
Protein G purified
Primary
Non-species specific
Purified
Western Blot, Immunohistochemistry
3G4-1F5
Western Blot, Immunohistochemistry
Gum levan, Kelacid, Levan gum, Sazio
Sodium Alginate conjugated to KLH
100 μL
Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA.
Store at 4C in the dark.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.