Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alginate Monoclonal Antibody (4B10-1C5), PerCP
GREENER_CHOICE

Catalog No. PIMA545313
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545313 Supplier Invitrogen™ Supplier No. MA545313
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45313 was sufficient for detection of 150 ng of alginate-conjugated BSA by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA.

Sodium alginate is used in biological experiments for the immobilization of cells and encapsulation due to its biocompatibility and simple gelation with divalent cations such as Ca2+. Studies suggest that preparation of alginate microspheres will sustain protein delivery within tissue scaffolds, as well as in many other cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alginate
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4B10-1C5
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene Alias Algin; Alginic Acid; Sodium Alginate
Host Species Mouse
Immunogen Sodium Alginate conjugated to KLH.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.