Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alginate Mouse anti-Human, FITC, Clone:4B10-1C5, Abnova™

Catalog No. 89333405 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-405 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-405 Supplier Abnova Corporation Supplier No. MAB16781
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Alginate.

Specifications

Antigen Alginate
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4B10-1C5
Conjugate FITC
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Host Species Mouse
Immunogen Sodium Alginate conjugated to KLH.
Quantity 100 μg
Target Species Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.