Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Alginic acid

Catalog No. AC270580025
Change view
Click to view available options
Quantity:
100 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270580025 2.5 kg Plastic bottle
AC270581000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270580025 Supplier Thermo Scientific Chemicals Supplier No. 270580025
Add to Cart

CAS: 9005-32-7 | (C6H8O7)A(C6H8O7)B

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-32-7
Molecular Formula (C6H8O7)A(C6H8O7)B
MDL Number MFCD00081309
Synonym 2r,3r,4r,5s,6s-3-2r,3s,4r,5s,6s-6-carboxy-3,4-dihydroxy-5-methoxyoxan-2-yl oxy-4,5-dihydroxy-6-methyloxane-2-carboxylic acid

Specifications

Melting Point >300°C
Color Beige to White
Ignition Residue 4% max.
Loss on Drying 15% max. (2 g, 105°C, 4 hrs)
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 2.5 kg
Merck Index 15,235
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in hot water;,insoluble in organic solvents;,dissolves slowly in solutions of sodium carbonate,,sodium hydroxide and trisodium phosphate
Percent Purity 19 to 25% (carboxyl groups) (on dried substance)
Physical Form Powder
Chemical Name or Material Alginic acid
Show More Show Less
EINECSNumber 232-680-1
RTECSNumber AZ5775000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.