Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Alginic acid sodium salt

Catalog No. 180947250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-094-7250G 250 g
18-094-7500G 500 g
18-094-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-094-7250G Supplier Sigma Aldrich Supplier No. 180947250G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alginic acid sodium is a gelling and nontoxic anionic polysaccharide. The carboxylic acid groups on the alginic acid chain, renders it insoluble in water.However, converting alginic acid to its sodium form, enables it to solubilize in water easily.

Alginic acid sodium is used:

  • In combination with chitosan, to fabricate a biodegradable porous scaffold for bone tissue engineering.
  • To study the characteristics of a modified amphiphilic alginate derivative. to the study the impact of alginate on the rate of lipid digestion by employing an in vitro digestion model .
  • In the preparation of alginate hydrogels.
  • As encapsulating agents of microparticles of β-galactosidae

Specifications

CAS 9005-38-3
Quantity 250 g
MDL Number MFCD00081310
Synonym Algin; Sodium alginate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.