Learn More
Aliquat™ 336TG
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.8900 g/mL |
| Flash Point | 77°C |
| Quantity | 1 L |
| Specific Gravity | 0.89 |
| Solubility Information | Solubility aq. soln.: insoluble. |
| Viscosity | 1500 mPa.s (30°C) |
| Chemical Name or Material | Aliquat* 336 TG |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May damage fertility. May damage the unborn child. Very toxic to aquatic l |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. |
| EINECSNumber | 264-120-7 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.