Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Aliskiren hemifumarate
GREENER_CHOICESDP

Catalog No. 549150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-915-0 50 mg
54-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 54-915-0 Supplier Tocris Bioscience™ Supplier No. 5491/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent renin inhibitor; antihypertensive

Aliskiren hemifumarate is a potent renin inhibitor (IC50 = 0.6 and 80 nM for human and rat respectively). Exhibits selectivity for renin over a range of other aspartic proteinases (>5000 nM). Lowers blood pressure in a hypertensive rodent model. Orally active.

Specifications

Quantity 50 mg
Product Type Aliskiren hemifumarate
Purity >99%
Molecular Weight (g/mol) 609.83

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.