Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alivin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562894 Supplier Invitrogen™ Supplier No. PA562894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQVGERLMVH CDSKTGNANT DFIWVGPDNR LLEPDKEMEN FYVFHNGSLV IESPRFEDAG VYSCIAMNKQ RLLNETVDVT INVSNFTVSR S Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 82%.

A novel primary response gene, Alivin 1 (ali1), is an activity-dependent gene and promotes survival of neurons. Sequence analyses reveal that rat, mouse, and human Ali1 proteins contain seven leucine-rich repeats, one IgC2-like loop and a transmembrane domain, and display homology to Kek and Trk families. Results suggest that expression of ali1 promotes depolarization-dependent survival of the granule neuron. Mouse ali1 was mapped to a locus approximately 55.3 cM from the centromere on chromosome 15 that is syntenic to positional candidate loci for familial Alzheimer's disease type 5 and Parkinson's disease 8 on human chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alivin 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Amigo2
Gene Accession No. Q86SJ2
Gene Alias adhesion molecule with Ig like domain 2; adhesion molecule with Ig-like domain 2; AI415330; Ali1; alivin 1; alivin-1; amigo2; AMIGO-2; amigo2 {ECO:0000312; amphoterin induced gene 2; amphoterin induced gene and ORF 2; amphoterin-induced protein 2; AW208913; DEGA; differentially expressed in gastric adenocarcinoma; differentially expressed in gastric adenocarcinomas; EMBL:AAO48951.1}; transmembrane protein AMIGO2
Gene Symbols Amigo2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Alivin 1. Recombinant protein control fragment (Product #RP-101320).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347902
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.