Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alix Antibody (3A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
$243.50 - $548.00
Specifications
Antigen | Alix |
---|---|
Clone | 3A9 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB165678SS
![]() |
Novus Biologicals
NB10065678SS |
0.025 mg |
Each for $243.50
|
|
|||||
NB10065678
![]() |
Novus Biologicals
NB10065678 |
0.1 mg |
Each for $548.00
|
|
|||||
Description
Alix Monoclonal specifically detects Alix in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Alix | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat, Xenopus species | |
AIP1DRIP4, ALG-2 interacting protein 1, ALG-2 interacting protein X, ALG-2-interacting protein 1, alinx, ALIX, apoptosis-linked gene 2-interacting protein X, dopamine receptor interacting protein 4, HP95, KIAA1375, MGC17003, PDCD6-interacting protein, programmed cell death 6 interacting protein, programmed cell death 6-interacting protein | |
PDCD6IP | |
IgG1 | |
Protein G purified | |
NB100-65678 recognizes apoptotic-linked-gene-product 2 (ALG-2) interacting protein X (ALIX), a conserved adaptor protein which is ubiquitously expressed. Alix was originally reported to play a role in apoptosis but has recently been shown to be involved in other cellular mechanisms including endosomal sorting, endocytosis, viral budding and actin cytoskeleton assembly. |
3A9 | |
Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Q8WUM4 | |
10015 | |
Alix fusion protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title