Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alix Antibody (7E9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Alix |
---|---|
Clone | 7E9 |
Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Alix Monoclonal antibody specifically detects Alix in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
Alix | |
Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
AIP1DRIP4, ALG-2 interacting protein 1, ALG-2 interacting protein X, ALG-2-interacting protein 1, alinx, ALIX, apoptosis-linked gene 2-interacting protein X, dopamine receptor interacting protein 4, HP95, KIAA1375, MGC17003, PDCD6-interacting protein, programmed cell death 6 interacting protein, programmed cell death 6-interacting protein | |
A synthesized peptide derived from Human Alix [UniProt Q8WUM4] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7E9 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
10015 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title