Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alix Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149701MFV500
Description
Alix Polyclonal antibody specifically detects Alix in Human, Mouse samples. It is validated for Flow CytometrySpecifications
Alix | |
Polyclonal | |
Flow Cytometry | |
AIP1DRIP4, ALG-2 interacting protein 1, ALG-2 interacting protein X, ALG-2-interacting protein 1, alinx, ALIX, apoptosis-linked gene 2-interacting protein X, dopamine receptor interacting protein 4, HP95, KIAA1375, MGC17003, PDCD6-interacting protein, programmed cell death 6 interacting protein, programmed cell death 6-interacting protein | |
The immunogen for this product maps to a region between residue 818 and 868 of human Apoptosis-Linked Gene 2-Interacting Protein X using the numbering given in entry NP_037506.2 (GeneID 10015). | |
0.1 mL | |
Apoptosis | |
10015 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction