Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alix Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103470
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103470 Supplier Invitrogen™ Supplier No. PA5103470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PDCD6IP.

Apoptotic linked-gene-product 2 (ALG-2) interacting protein X (ALIX) is a conserved adaptor protein which is ubiquitously expressed. Alix was originally reported to play a role in apoptosis but has recently been shown to be involved in other cellular mechanisms including endosomal sorting, endocytosis, viral budding and actin cytoskeleton assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alix
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Pdcd6ip
Gene Accession No. Q8WUM4, Q9QZA2, Q9WU78
Gene Alias AI480591; AIP1; ALG-2 interacting protein 1; ALG-2 interacting protein X; ALG-2-interacting protein 1; Alg2-interacting protein 1; ALG-2-interacting protein X; Alg2-interacting protein X; Alix; apoptosis-linked gene 2-interacting protein X; AW544830; C76364; dopamine receptor interacting protein 4; DRIP4; E2F1-inducible protein; Eig2; Hp95; KIAA1375; MGC17003; mKIAA1375; PDCD6-interacting protein; Pdcd6ip; pdcd6ip.L; programmed cell death 6 interacting protein; programmed cell death 6 interacting protein L homeolog; programmed cell death 6-interacting protein; putative signal tranduction protein Xp95; RGD1561176; Signal transduction protein Xp95; XELAEV_18031317mg; xp95
Gene Symbols Pdcd6ip
Host Species Rabbit
Immunogen A synthesized peptide derived from human PDCD6IP(Accession Q8WUM4), corresponding to amino acid residues I89-N139.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10015, 18571, 501083
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.