Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alix Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$201.00 - $584.50
Specifications
Antigen | Alix |
---|---|
Concentration | 1.0 mg/mL |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Alix Polyclonal antibody specifically detects Alix in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin).Specifications
Alix | |
Polyclonal | |
Rabbit | |
Apoptosis | |
AIP1DRIP4, ALG-2 interacting protein 1, ALG-2 interacting protein X, ALG-2-interacting protein 1, alinx, ALIX, apoptosis-linked gene 2-interacting protein X, dopamine receptor interacting protein 4, HP95, KIAA1375, MGC17003, PDCD6-interacting protein, programmed cell death 6 interacting protein, programmed cell death 6-interacting protein | |
PDCD6IP | |
IgG | |
Affinity Purified |
1.0 mg/mL | |
Unconjugated | |
RUO | |
PBS with 0.09% Sodium Azide | |
10015 | |
The immunogen for this product maps to a region between residue 818 and 868 of human Apoptosis-Linked Gene 2-Interacting Protein X using the numbering given in entry NP_037506.2 (GeneID 10015). | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title