Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin Complexone, may cont. up to 10% water

Catalog No. AAA1669903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1669903 1 g
AAA1669906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1669903 Supplier Thermo Scientific Chemicals Supplier No. A1669903
Add to Cart

CAS: 3952-78-1 | C19H15NO8 | 385.328 g/mol

Alizarin is the main ingredient for the manufacture of the madder lake pigments known to painters as Rose madder and Alizarin crimson. A notable use of alizarin in modern times is as a staining agent in biological research because it stains free calcium and certain calcium compounds a red or light purple color. Alizarin Red is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alizarin is the main ingredient for the manufacture of the madder lake pigments known to painters as Rose madder and Alizarin crimson. A notable use of alizarin in modern times is as a staining agent in biological research because it stains free calcium and certain calcium compounds a red or light purple color. Alizarin Red is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3952-78-1
Molecular Formula C19H15NO8
Molecular Weight (g/mol) 385.328
MDL Number MFCD00001202
InChI Key PWIGYBONXWGOQE-UHFFFAOYSA-N
Synonym Alizarin Fluorine Blue; Alizarin-3-methyliminodiacetic acid
PubChem CID 65132
ChEBI CHEBI:53088
IUPAC Name 2-[carboxymethyl-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl]amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Melting Point 180°C to 182°C
Beilstein 2190028
Solubility Information Soluble in water
Formula Weight 385.33
Quantity 1 g
Assay may cont. up to 10% water
Chemical Name or Material Alizarin Complexone
EINECSNumber 223-544-2
RTECSNumber AH0585000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.