Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alizarin Red S Indicator, 0.1% w/v CI 58005, Reagents
Click to view available options
Quantity:
120 mL
Specifications
Specifications
Chemical Name or Material | Alizarin Red S Indicator, 0.1% w/v CI 58005 |
CAS | 7732-18-5, 130-22-3 |
Boiling Point | 100°C |
Percent Purity | 0.1% |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1.0001 |
Color | Orange/Red |
Quantity | 120 mL |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction