Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alizarin Red S Indicator, 1% w/v, Reagents
Click to view available options
Quantity:
120 mL
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Alizarin Red S Indicator, 1% w/v |
CAS | 7732-18-5, 130-22-3 |
Boiling Point | 100°C |
Percent Purity | 1% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.001 |
Color | Dark Red |
pH | 4.5 to 5 |
Quantity | 500 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction