Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin Red S Indicator, 1% w/v, Reagents

Catalog No. 01336059
Click to view available options
Quantity:
120 mL
500 mL
1 L

Alizarin Red S Indicator, 1% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Alizarin Red S Indicator, 1% w/v
CAS 7732-18-5, 130-22-3
Boiling Point 100°C
Percent Purity 1%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.001
Color Dark Red
pH 4.5 to 5
Quantity 120 mL
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.