Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Alizarin Red S, pure, certified

Catalog No. p-51926
Click to view available options
Quantity:
25 g
100 g

CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-22-3
Molecular Formula C14H7NaO7S
Molecular Weight (g/mol) 342.253
MDL Number MFCD00013049
InChI Key HFVAFDPGUJEFBQ-UHFFFAOYSA-M
Synonym 9,10-Dihydro-3,4-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid, sodium salt, 9, 10-Dihydro-3, 4-dihydroxy-9, Mordant Red 3, Sodium alizarinesulfonate
PubChem CID 3955344
ChEBI CHEBI:87358
IUPAC Name sodium;3,4-dihydroxy-9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=CC(=C(C(=C3C2=O)O)O)S(=O)(=O)[O-].[Na+]

Specifications

Absorption Ratio 0.97 to 1.07 (for the first maximum) (P-15/P+15)
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in ethanol
Infrared Spectrum Authentic
Lambda Max 554 to 558nm 590 to 599nm
Formula Weight 342.26
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 3.5 (yellow) to pH 6.5 (pink) and from pH 9.4 (orange-brown) to pH 12.0 (violet)
Color Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Alizarin Red S
Show More Show Less
Hazard Category Warning
EINECSNumber 204-981-8
RTECSNumber CB1095300
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.