Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALK-1 Antibody (104), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289256JF646
Description
ALK-1 Monoclonal antibody specifically detects ALK-1 in Human samples. It is validated for ELISA, ELISASpecifications
ALK-1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
104 | |
ELISA, Sandwich ELISA | |
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ALK-1 (NP_000011.2; Met1-Gln118). | |
0.1 mL | |
Protein Kinase | |
94 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction