Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALK/CD246 Antibody (ALK/1032) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308772
Description
ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Western Blot.Specifications
ALK/CD246 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL | |
ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 | |
Recombinant fragment (around aa 220-335) of human ALK/CD246 protein (exact sequence is proprietary) | |
100 μg | |
Phospho Specific, Protein Kinase, Signal Transduction | |
238 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot | |
ALK/1032 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction