Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALK/CD246 Antibody (ALK1/2766R), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308447C
Description
ALK/CD246 Monoclonal antibody specifically detects ALK/CD246 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ALK/CD246 | |
Monoclonal | |
DyLight 650 | |
ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 | |
Recombinant full-length human ALK/CD246 protein (Uniprot: Q9UM73) | |
100 μg | |
Protein Kinase, Signal Transduction | |
238 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
ALK1/2766R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction