Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK, Mouse anti-Human, Clone: ALK1, Millipore Sigma™

Catalog No. MABC117125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC117125 25 μg
MABC1171100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC117125 Supplier MilliporeSigma Supplier No. MABC117125UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-ALK, clone ALK1, Cat. No. MABC1171, is a mouse monoclonal antibody that detects ALK tyrosine kinase receptor and has been tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

ALK tyrosine kinase receptor (UniProt: Q9UM73; also known as EC: 2.7.10.1, Anaplastic lymphoma kinase, CD246) is encoded by the ALK gene (Gene ID: 238) in human. ALK tyrosine kinase receptor is a single-pass type I membrane protein that is essentially and transiently expressed in specific regions of the central and peripheral nervous systems and plays an important role in the development and function of the nervous system. It is predominantly expressed in the central nervous system, intestine, and testis. It acts as a receptor for ligands pleiotrophin (PTN) and midkine (MDK). It is activated and homodimerizes by ligand-binding and subsequent phosphorylation. Following activation by ligand, ALK induces tyrosine phosphorylation of CBL, FRS2, IRS1 and SHC1, as well as of the MAP kinases MAPK1/ERK2 and MAPK3/ERK1. It is inactivated through dephosphorylation by receptor protein tyrosine phosphatase beta and zeta complex (PTPRB/PTPRZ1) in the absence of any stimulation by a ligand. A chromosomal aberration involving ALK has been reported in non-Hodgkin lymphoma where the kinase is constitutively activated. Another chromosomal aberration involving ALK has also been linked with anaplastic large-cell lymphoma (ALCL). ALK tyrosine kinase receptor is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off in the mature form. ALK tyrosine kinase receptor consists of an extracellular domain (aa 19-1038); a transmembrane domain (aa 1039-1059); and a cytoplasmic domain (aa 1060-1620).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALK
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone ALK1
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected ALK in Western Blotting applications (Pulford, K., et. al. (2002). Br J Haematol. 116(1):135-41; Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunohistochemistry Analysis: A representative lot detected ALK in Immunohistochemistry applications (Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunofluorescence Analysis: A representative lot detected ALK in Immunofluorescence applications (Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunoprecipitation Analysis: A representative lot immunoprecipitated ALK in Immunoprecipitation applications (Pulford, K., et. al. (2002). Br J Haematol. 116(1):135-41; Pulford, K., et. al. (1997). Blood. 89(4):1394-404).Immunohistochemistry (Paraffin) Analysis: A representative lot detected ALK in Immunohistochemistry applications (Pulford, K., et. al. (1997). Blood. 89(4):1394-404).
Formulation Purified mouse monoclonal antibody IgG3 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias ALK tyrosine kinase receptor;EC: 2.7.10.1;Anaplastic lymphoma kinase;CD246
Gene Symbols ALK
Host Species Mouse
Immunogen His-tagged full length human recombinant ALK tyrosine kinase receptor.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_004295
Test Specificity Clone ALK1 specifically detects human ALK tyrosine kinase receptor. It targets an epitope with in 102 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.