Learn More
ALK Mouse anti-Human, Clone: OTI1E12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800730

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas. The translocation creates a fusion gene consisting of the ALK gene and the nucleophosmin gene: the 3′ half of ALK, derived from chromosome 2, is fused to the 5′ portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK. ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system.Specifications
ALK | |
Monoclonal | |
Unconjugated | |
ALK | |
CD246, NBLST3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
238 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1E12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UM73 | |
ALK | |
Full length human recombit protein of human ALK produced in HEK293T cell | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.