Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALK Mouse anti-Human, Unconjugated, Clone: 2A5, Abnova™

Catalog No. 89335154 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-154 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-154 Supplier Abnova Corporation Supplier No. H00000238M06
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ALK Monoclonal antibody specifically detects ALK in Human samples. It is validated for ELISA.

Specifications

Antigen ALK
Applications ELISA
Classification Monoclonal
Clone 2A5
Conjugate Unconjugated
Description anaplastic lymphoma receptor tyrosine kinase
Gene ALK
Gene Accession No. BC156207.1
Gene Alias CD246, Ki-1, TFG/ALK
Host Species Mouse
Immunogen A synthetic peptide corresponding to human ALK
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 238
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.