Learn More
ALK (phospho Y1604) Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human ALK.
Supplier: Abnova Corporation PAB29642
Description
The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas (ALCLs). The translocation creates a fusion gene consisting of the ALK (anaplastic lymphoma kinase) gene and the nucleophosmin (NPM) gene: the 3' half of ALK, derived from chromosome 2, is fused to the 5' portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK (leukocyte tyrosine kinase). ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system. [provided by RefSeq]
Specifications
ALK (phospho Y1604) | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human ALK. | |
In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
ALK | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y1604 of human ALK. | |
100 μL | |
Primary | |
ALK (phospho Y1604) polyclonal antibody detects endogenous levels of human ALK only when phosphorylated at tyrosine 1604. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:100-1:500) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
CD246/Ki-1/TFG/ALK | |
Rabbit | |
Affinity chromatography | |
RUO | |
238 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.