Learn More
Promega ALK2 Kinase Enzyme System, 10microg
Catalog No. PRV4492 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4492 10 μg
1 options
Catalog No. PRV4492 Supplier Promega Supplier No. V4492
ALK2 is a receptor serine/threonine kinase that is member of the ALK family and is upstream of signaling pathway involving the SMAD proteins especially SMAD1/5/8.
  • Easily Screen and Profile ALK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C